missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KCNQ2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 11538611
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11538611 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11538611 Supplier Invitrogen™ Supplier No. PA1929

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PA1-929 detects KCNQ2 from mouse, human and rat samples. This antibody is specific for KCNQ2 and does not detect KCNQ1, KCNQ3, KCNQ4 or KCNQ5. PA1-929 has been successfully used in Western blot, immunofluorescence, immunohistochemistry and immunocytochemistry procedures. Immunohistochemical staining of rat hippocampal using PA1-929 yielded a strong signal in granule cell layer and the mossy fibers found in the central hilus of the dentate gyrus. PA1-929 immunogen is a GST fusion protein encoding the first 70 amino acids of human KCNQ2.

KCNQs are members of the voltage-dependent non-inactivating potassium channel family. Currently there are five known KNCQs (KCNQ1-5) found in the central nervous system and KNCQ2 and 3 have demonstrated their importance in M-current activation. Studies have shown that KCNQ2 and KCNQ3 form heteromultimers that, when formed, substantially increase the M-current. Inhibition of M-current controls neuron excitability throughout the nervous system as well as the responsiveness to synaptic inputs. Genetic mutations in these proteins have been linked to disorders such as benign familial neonatal convulsions (BFNC), deafness, neuropathic pain and epilepsy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNQ2
Applications Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.02% sodium azide
Gene KCNQ2
Gene Accession No. O43526, O88943, Q9Z351
Gene Alias BFNC; EBN; EBN1; ENB1; HNSPC; KCNA11; KCNQ2; KQT like 2; KQT2; KQT-like 2; KV7.2; KVEBN1; mKQT2.3; mKQT2.4; neuroblastoma-specific potassium channel subunit alpha KvLQT2; Nmf134; potassium channel subunit alpha KvLQT2; potassium channel, voltage gated KQT-like subfamily Q, member 2; potassium channel, voltage-gated KQT-like subfamily Q, member 2; potassium voltage-gated channel KQT-like subfamily member 2.3; potassium voltage-gated channel KQT-like subfamily member 2.4; potassium voltage-gated channel subfamily KQT member 2; potassium voltage-gated channel subfamily Q member 2; potassium voltage-gated channel, KQT-like subfamily, member 2; potassium voltage-gated channel, subfamily Q, member 2; voltage-gated potassium channel subunit Kv7.2
Gene Symbols KCNQ2
Host Species Rabbit
Immunogen GST fusion protein encoding the first 70 amino acids of human KCNQ2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16536, 170848, 3785
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.