missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNMA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577623
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
Potassium channels are a group of ubiquitously expressed proteins that serve numerous functions in excitable and non-excitable cells. One class of integral membrane potassium channels is the large conductance, calcium-activated potassium channel (Maxi K+). Maxi K+ differs from most other potassium channels in that its activation is controlled by both increases in intracellular calcium and by membrane depolarization. Maxi K+ dual activation is possible because of its structure. The core of the channel, which is similar to other potassium channels, is a Maxi K+ alpha homotetramer that contains both a voltage sensor and an intracellular calcium binding domain. In vascular smooth muscle, an auxiliary beta-subunit is found in a 1:1 stoichiometry. The beta-subunit exhibits its effect on the Maxi K+ channel by effectively decreasing by 5- to 10- fold the concentration of calcium required to keep the pore open.
Specifications
| KCNMA1 | |
| Polyclonal | |
| Unconjugated | |
| KCNMA1 | |
| BKCa, KCNMA1, SLO | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16531, 3778, 83731 | |
| -20°C | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.6 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| Q08460, Q12791, Q62976 | |
| Kcnma1 | |
| Peptide (C)STANRPNRPKSRESRDK, corresponding to amino acid residues 1184-1200 of mouse KCa 1.1 | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction