missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCNH5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541073
This item is not returnable.
View return policy
Description
Peptide sequence: LTNSRSVLQQ LTPMNKTEVV HKHSRLAEVL QLGSDILPQY KQEAPKTPPH Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. The KCNH5 gene encodes a member of the potassium channel, voltage-gated, subfamily H. This member is a pore-forming (alpha) subunit of a voltage-gated non-inactivating delayed rectifier potassium channel. KCNH5 is not expressed in differentiating myoblasts.
Specifications
| KCNH5 | |
| Polyclonal | |
| Unconjugated | |
| KCNH5 | |
| EAG2; Ether-a-go-go potassium channel 2; ether-a-go-go-related potassium channel 2; hEAG2; H-EAG2; KCNH5; Kv10.2; potassium channel, voltage gated eag related subfamily H, member 5; potassium voltage-gated channel subfamily H member 5; potassium voltage-gated channel, subfamily H (eag-related), member 5; rEAG2; Voltage-gated potassium channel subunit Kv10.2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 27133 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8NCM2 | |
| KCNH5 | |
| Synthetic peptide directed towards the N-terminal of human KCNH5 (aa 156-205). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction