missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCNH2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568667
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 86%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 93%; Rabbit: 100%; Rat: 100%.
Human ether-a-go-go related gene (HERG) encodes the pore-forming alpha subunit of the delayed rectifier potassium channel IKr. There are two N-terminal splice variants of HERG include the full-length isoform 1 alpha and the shorter isoform 1 beta. Isoform 1 beta lacks the PAS motif and deactivates at a faster rate than isoform 1alpha. Residues within the C-terminal play a role in channel expression and channel gating, including voltage-dependent activation. HERG is expressed in the heart and is more abundantly expressed in the ventricles than in the atria. Mutations in the gene encoding HERG increase beat-to-beat variability and early after depolarization. These fluctuations facilitate the genesis and propagation of premature heartbeats associated with inheritable long QT syndrome type 2 and short QT syndrome type 1.
Specifications
| KCNH2 | |
| Polyclonal | |
| Unconjugated | |
| KCNH2 | |
| AI326795; cerg; c-ERG; cERG; ether-a-go-go related member 2; delayed rectifier potassium channel; derg; Eag homolog; eag related protein 1; eag-related gene; eag-related protein 1; ERG; ERG1; ERG-1; ether-a-go-go related gene potassium channel 1; ether-a-go-go related protein; ether-a-go-go-related gene potassium channel 1; ether-a-go-go-related potassium channel protein; ether-a-go-go-related protein 1; gp-erg; HERG; H-ERG; HERG1; HERG-1; KCNH2; Kv11.1; LQT; Lqt2; M-erg; Merg1; M-erg1; merg1a; merg1b; potassium channel; potassium channel protein ERG; potassium channel protein ERG long isoform; potassium channel protein ERG short isoform; potassium channel, voltage gated eag related subfamily H, member 2; potassium voltage-gated channel subfamily H member 2; potassium voltage-gated channel, subfamily H (eag-related), member 2; ra-erg; RERG; r-ERG; SQT1; ventricular ERG K+ channel subunit; voltage-gated potassium channel 11.1; voltage-gated potassium channel subunit Kv11.1; voltage-gated potassium channel variant 3; voltage-gated potassium channel, subfamily H, member 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 16511 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O35219 | |
| KCNH2 | |
| synthetic peptide directed towards the middle region of human KCNH2 (aa 895-944). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction