missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KBTBD7 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15748311 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15748311 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15748311 Supplier Invitrogen™ Supplier No. PA560478

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: SDDEVWVQVA PQRNAQDQQG SL Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 41%.

The protein encoded by this gene is a transcriptional activator, having been shown to increase the transcription of activator protein-1 and serum response element. The encoded protein can also form a complex with KBTBD6 and CUL3, which regulates the ubiquitylation and degradation of TIAM1, which is a regulator of RAC1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KBTBD7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KBTBD7
Gene Accession No. Q8WVZ9
Gene Alias KBTBD7; kelch repeat and BTB (POZ) domain containing 7; kelch repeat and BTB domain containing 7; kelch repeat and BTB domain-containing protein 7; kelch repeat and BTB domain-containing protein 7-like
Gene Symbols KBTBD7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KBTBD7. Recombinant protein control fragment (Product #RP-99579).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84078
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.