missing translation for 'onlineSavingsMsg'
Learn More

KBTBD2 Mouse Monoclonal Antibody, Proteintech

Product Code. 30235527 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30235527 150 μL 150µL
30234202 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30235527 Supplier Proteintech Supplier No. 678951IG150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The KBTBD2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human KBTBD2. This antibody recognizes human, mouse, rat antigen. The KBTBD2 antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.

Specifications

Antigen KBTBD2
Applications Western Blot, Immunocytochemistry/Immunofluorescence, ELISA
Classification Monoclonal
Clone 2A9F3
Concentration 1000 μg/mL
Conjugate Unconjugated
Gene Symbols BKLHD1
Host Species Mouse
Immunogen Recombinant Protein
Purification Method Protein G purification
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25948
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 60-70 kDa
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.