Learn More
kb-NB77-78, MedChemExpress
Product Code. 16419677
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16419677 5 mg 5mg
16477138 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16419677 Supplier Medchem Express Supplier No. HY16698
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
MedChemExpress kb-NB77-78 is an analogue of CID797718, but shows no PKD inhibitory activity.
kb-NB77-78 is an analogue of CID797718, but shows no PKD inhibitory activity.
Specifications
Chemical Name or Material kb-NB77-78
CAS 1350622-33-1
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C18H25NO3Si
Quantity 5 mg
Solubility Information DMSO : 25 mg/mL (75.42 mM; Need ultrasonic)
SMILES O=C1OC2=CC=C(O[Si](C)(C(C)(C)C)C)C=C2C3=C1NCCC3
Molecular Weight (g/mol) 331.48
Formula Weight 331.48
Percent Purity 99.97%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death