missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kappa Light Chain Antibody (A8B5), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-45067IR
This item is not returnable.
View return policy
Description
Kappa Light Chain Monoclonal specifically detects Kappa Light Chain in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Kappa Light Chain | |
| Monoclonal | |
| DyLight 755 | |
| HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| The antibody A8B5 reacts with IGKC (22.5 kDa) of immunoglobulins. Cross reacts with Human. Does not cross react with Hamster, Goat, Guinea Pig, Rabbit and Sheep. | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| A8B5 | |
| Flow Cytometry, ELISA, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| IGKC | |
| The antibody A8B5 reacts withIGKC (22.5 kDa) of immunoglobulins. | |
| 0.1 mL | |
| Adaptive Immunity, Immunology | |
| 3514 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction