Learn More
Kanamycin sulfate, MedChemExpress
Product Code. 16419627
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16419627 5 g 5g
16471619 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16419627 Supplier Medchem Express Supplier No. HY16566A
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
MedChemExpress Kanamycin (Kanamycin A) sulfate is an orally active antibacterial (gram-negative/positive bacteria) agent, inhibits translocation and causes misencoding by binding to the 70 S ribosomal subunit. Kanamycin sulfate shows good inhibitory activity to both M. tuberculosis (sensitive and drug-resistant ) and K. pneumonia, which can be used in studies of tuberculosis and pneumonia.
Kanamycin (Kanamycin A) sulfate is an orally active antibacterial (gram-negative/positive bacteria) agent, inhibits translocation and causes misencoding by binding to the 70 S ribosomal subunit. Kanamycin sulfate shows good inhibitory activity to both M. tuberculosis (sensitive and drug-resistant ) and K. pneumonia, which can be used in studies of tuberculosis and pneumonia.
Specifications
Chemical Name or Material Kanamycin sulfate
Physical Form Solid
CAS 25389-94-0
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C18H38N4O15S
For Use With (Application) COVID-19-immunoregulation
Synonym Kanamycin A sulfate
Solubility Information H2O : 31.25 mg/mL (53.64 mM; ultrasonic and warming and heat to 80°C) ∣DMSO : 1 mg/mL (1.72 mM; ultrasonic and warming and heat to 80°C)
SMILES OS(O)(=O)=O.O[C@H]1[C@](O[C@@H]2[C@@H](N)C[C@@H](N)[C@H](O[C@@]3([H])O[C@H](CO)[C@@H](O)[C@H](N)[C@H]3O)[C@H]2O)([H])O[C@H](CN)[C@@H](O)[C@@H]1O
Molecular Weight (g/mol) 582.58
Formula Weight 582.58
Percent Purity 95.0%
Grade Research