missing translation for 'onlineSavingsMsg'
Learn More

Kainic acid, MedChemExpress

Product Code. p-200100582
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
This item is not returnable. View return policy

Product Code. 16469278

Brand: Medchem Express HYN2309

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Kainic acid is a potent agonist at excitatory amino acid receptor subtypes in the CNS.

Kainic acid is a potent agonist at excitatory amino acid receptor subtypes in the CNS.

Specifications

Chemical Name or Material Kainic acid
CAS 487-79-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C10H15NO4
Quantity 10 mg
Solubility Information H2O : 25 mg/mL (117.24 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(O)C[C@@H]1[C@@H](C(O)=O)NC[C@@H]1C(C)=C
Molecular Weight (g/mol) 213.23
Formula Weight 213.23
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.