missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ kaiC Polyclonal Antibody
Description
kaiC Polyclonal Antibody for Western Blot, ELISA
Central component of the KaiABC oscillator complex, which constitutes the main circadian regulator in cyanobacteria. Complex composition changes during the circadian cycle to control KaiC phosphorylation. KaiA stimulates KaiC autophosphorylation, while KaiB sequesters KaiA, leading to KaiC autodephosphorylation. Clock output pathways impact the RpaA transcriptional regulator. KaiC enhances the autophosphorylation activity of SasA, which then transfers its phosphate group to RpaA to activate it. KaiB and KaiC together enhance the phospho-RpaA dephosphatase activity of CikA.
Specifications
Specifications
| Antigen | kaiC |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 3.69 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 |
| Gene Alias | Circadian clock protein kinase KaiC; kaiC |
| Host Species | Rabbit |
| Immunogen | Recombinant Nostoc sp. (strain PCC 7120/SAG 25.82/UTEX 2576) Circadian clock protein kinase KaiC (1-270aa). |
| Purification Method | Protein G |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?