missing translation for 'onlineSavingsMsg'
Learn More

K-80003, MedChemExpress

Product Code. p-200097259
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490459 10 mg 10mg
16402668 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16490459 Supplier Medchem Express Supplier No. HYU00458

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress K-80003 is a potent inhibitor of tRXRα-dependent Akt activation and cancer cell growth.

K-80003 is a potent inhibitor of tRXRα-dependent Akt activation and cancer cell growth.

Specifications

Chemical Name or Material K-80003
CAS 1292821-90-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H21FO2
Quantity 10 mg
Synonym TX-803
Solubility Information DMSO : 30 mg/mL (89.18 mM; Need ultrasonic)
SMILES CC1=C(C2=CC(F)=CC=C2/C1=C(C3=CC=C(C=C3)C(C)C)/[H])CC(O)=O
Molecular Weight (g/mol) 336.4
Formula Weight 336.4
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.