Learn More
K-80003, MedChemExpress
Product Code. 16490459
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16490459 10 mg 10mg
16402668 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16490459 Supplier Medchem Express Supplier No. HYU00458
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress K-80003 is a potent inhibitor of tRXRα-dependent Akt activation and cancer cell growth.
K-80003 is a potent inhibitor of tRXRα-dependent Akt activation and cancer cell growth.
Specifications
Chemical Name or Material K-80003
CAS 1292821-90-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H21FO2
Quantity 10 mg
Synonym TX-803
Solubility Information DMSO : 30 mg/mL (89.18 mM; Need ultrasonic)
SMILES CC1=C(C2=CC(F)=CC=C2/C1=C(C3=CC=C(C=C3)C(C)C)/[H])CC(O)=O
Molecular Weight (g/mol) 336.4
Formula Weight 336.4
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease