Learn More
Justrite™ Sure-Grip™ EX Flammable Safety Cabinet
Sure-Grip™ EX Flammable Safety Cabinet protects workers, reduces fire risks, and improves worker productivity by allowing the storage of flammable fuels and chemicals in code-compliant, 18-gauge steel safety cabinets.
1232.35 €
Specifications
| Color | Yellow |
|---|---|
| Product Type | Can storage cabinet |
| For Use With (Application) | Flammable liquid storage and protection |
| Includes | U-Loc™ padlockable handle, Haz-Alert™ reflective trilingual warning labels, sturdy SpillSlope™ shelves, three-point Stainless Steel bullet self latching system and welded construction, dual vents with |
| Material | Steel (CR) |
| Product Code | Brand | Load Bearing Capacity | Dimensions (D x W x H) Interior | Door Style | Weight (Metric) | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Load Bearing Capacity | Dimensions (D x W x H) Interior | Door Style | Weight (Metric) | Price | Quantity & Availability | ||
|
11945346
|
Justrite™
8930201 |
30 gal. | 14.56 x 39.5 x 39.62 in. (37 x 100.3 x 100.6 cm) | Self-close | 176 kg |
1585.36 € 1232.35 € / Pack of 1 Save 353.01 € 22% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Sure-Grip™ EX Flammable Safety Cabinet protects workers, reduces fire risks, and improves worker productivity by allowing the storage of flammable fuels and chemicals in code-compliant, 18-gauge steel safety cabinets.
- Sure-Grip EX flush-mounted door handle does not interfere with traffic; comes with two keys for security and can be padlocked (padlock not included)
- Exclusive self-latching feature ensures that cabinet doors are safely closed without manually engaging door handle
- Double-wall construction provides 1.5 in. (3.8cm) of insulating air space to help prevent spread of flames High-visibility Haz-Alert™ reflective warning labels offer added safety during power outages or fires
- Dual vents with flame arresters at top and opposite bottom allow hazardous vapors to be piped away Easy-closing, self-latching doors facilitate fire protection of the cabinet
- Self-latch engages critical three-point latching system for protection under fire conditions
- Fusible links hold self-closing doors wide open, flush with front of cabinet for easy access and resupply; automatically release and slam doors shut at 74°C (165°F)
- Concealed self-close mechanism provides optimum storage space and obstruction-free access to contents
- Two-door models hinged left and right have self-closing mechanism that ensures proper closing sequence regardless of which door is released first
- SpillSlope™ safety shelves direct incidental spills to the back and bottom of leakproof sump
- Adjustable leveling feet ensure solid positioning and alignment for tight door closure
- Exterior side panel has built-in grounding connector for easy grounding
- Finished inside and out with durable lead-free powder paint
- Meet OSHA and NFPA requirements
Certifications
FM approved except 19-120-2336.
Compliance
Meet NFPA Code 30 and OSHA regulations; models with self-close doors also comply with the Uniform Fire Code.
Warranty
10-Year
Specifications
| Yellow | |
| Flammable liquid storage and protection | |
| Steel (CR) | |
| Epoxy, Polyester |
| Can storage cabinet | |
| U-Loc™ padlockable handle, Haz-Alert™ reflective trilingual warning labels, sturdy SpillSlope™ shelves, three-point Stainless Steel bullet self latching system and welded construction, dual vents with | |
| 2 |