missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jumonji/JARID2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
195.00 € - 463.00 €
Specifications
| Antigen | Jumonji/JARID2 |
|---|---|
| Concentration | 1.0 mg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18389151
|
Bio-Techne
NBP3-00588 |
0.1 mg |
463.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18699517
|
Novus Biologicals
NBP3-00588SS |
0.025 mg |
195.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Jumonji/JARID2 Polyclonal antibody specifically detects Jumonji/JARID2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Jumonji/JARID2 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| JMJjumonji-like protein, jumonji (mouse) homolog, jumonji homolog, jumonji, AT rich interactive domain 2, Jumonji/ARID domain-containing protein 2, protein Jumonji | |
| Partial recombinant protein made to the N-terminal portion of the human Jumanji protein (amino acids 1-159) [UniProt Q92833] | |
| Immunogen affinity purified |
| 1.0 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Chromatin Research, Epigenetics, Neuroscience, Transcription Factors and Regulators | |
| PBS | |
| 3720 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title