missing translation for 'onlineSavingsMsg'
Learn More

JTV1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16866803 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16866803 20 μL 20µL
16856803 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16866803 Supplier Proteintech Supplier No. 104241AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This is a rabbit polyclonal antibody raised against full-length of JTV1.

The JTV1 gene is located on chromosome 7p22 flanked by two genes, HRI and PMS2. JTV1 and HRI overlap slightly and are arranged in a tail-to-tail fashion. JTV1 and PMS2 are separated by approximately 200 base pairs and are arranged head-to-head. JTV1 is transcribed in the opposite direction compared to HRI and PMS2. The function of the JTV1 gene product is unknown.

Specifications

Antigen JTV1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene AIMP2
Gene Accession No. Q13155, Q32PX2, Q8R010
Gene Alias AIMP2, JTV1, JTV1 gene, P38, PRO0992, Protein JTV 1
Gene Symbols Aimp2
Host Species Rabbit
Immunogen JTV1 Fusion Protein Ag0675
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 231872, 288480, 7965
Target Species Rat, Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.