missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ JPH2 Antibody, Novus Biologicals™

Product Code. 30041792 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30041792 50 μL 50µL
30041508 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30041792 Supplier Novus Biologicals™ Supplier No. NBP32786250ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

JPH2 Polyclonal antibody specifically detects JPH2 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen JPH2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-2000, ELISA 1:5000-20000
Formulation PBS, 50% glycerol
Gene Alias JP2, junctophilin 2, junctophilin type 2, junctophilin-2
Host Species Rabbit
Immunogen Synthesized peptide derived from human protein (Uniprot #: Q9BR39)
Purification Method Antigen Affinity-purified
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57158
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.