Learn More
Eurofins DiscoverX JNK1α1/SAPK1c Protein, Unactive
Description
c-Jun N-terminal kinases (JNKs), a stress-activated protein kinase (SAPK), is involved in cellular responses to environmental stresses. JNK kinases can be activated by stimuli such as UV light, radiation, protein synthesis inhibitors, ceramide, DNA-damaging drugs, chemopreventive drugs, TNF-a, and interleukin 1. JNK binds to an NH2-terminal region of ATF2 and c-Jun and phosphorylates two sites within the activation domain of each transcription factor. Three JNK genes (Jnk1, -2 and -3) have been identified in humans; however, splice variants result in a total of 10 isoforms. Therapeutic inhibition of JNK may provide clinical benefit in disease as diverse as immune inflammatory (joint, bowel, pulmonary), ischemic injury (brain, cardiac) and neurodegenerative (Alzheimer's, Parkinson's) diseases.
EMBL L26318
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 0.1 mM EGTA, 150 mM NaCl, 0.03% Brij-35, 50% glycerol, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptothanol. Liquid at -20°C. |
| Molecular Weight (g/mol) | 45 kDa |
| Quantity | 50 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Full-length |
| Purity or Quality Grade | ≥69% |
| Protein | JNK1a1, unactive |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.