Learn More
JNK-IN-8, MedChemExpress
Product Code. 16495638
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16495638
Brand: Medchem Express HY13319
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress JNK-IN-8 (JNK Inhibitor XVI) is a potent JNK inhibitor with IC50s of 4.7 nM, 18.7 nM, and 1 nM for JNK1, JNK2, and JNK3, respectively.
JNK-IN-8 (JNK Inhibitor XVI) is a potent JNK inhibitor with IC50s of 4.7 nM, 18.7 nM, and 1 nM for JNK1, JNK2, and JNK3, respectively.
Specifications
Chemical Name or Material JNK-IN-8
Physical Form Solid
CAS 1410880-22-6
Color Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C29H29N7O2
For Use With (Application) Cancer-Kinase/protease
Synonym JNK Inhibitor XVI
Solubility Information DMSO : ≥ 35 mg/mL (68.95 mM)
SMILES CC1=CC(NC(C2=CC=CC(NC(/C=C/CN(C)C)=O)=C2)=O)=CC=C1NC3=NC=CC(C4=CN=CC=C4)=N3
Molecular Weight (g/mol) 507.59
Formula Weight 507.59
Percent Purity 99.59%
Grade Research