Learn More
JNJ-42153605, MedChemExpress
Product Code. 16489947
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16489947 5 mg 5mg
16477468 10 mg 10mg
16447436 100 mg 100mg
16423497 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16489947 Supplier Medchem Express Supplier No. HY18162
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
MedChemExpress JNJ-42153605 is a positive allosteric modulator of the metabotropic glutamate 2 (mGlu2) receptor with an EC50 of 17 nM.
JNJ-42153605 is a positive allosteric modulator of the metabotropic glutamate 2 (mGlu2) receptor with an EC50 of 17 nM.
Specifications
Chemical Name or Material JNJ-42153605
CAS 1254977-87-1
Color White
Purity Grade Notes Research
Molecular Formula C22H23F3N4
Quantity 5 mg
Solubility Information DMSO : 16.67 mg/mL (41.63 mM; Need ultrasonic)
SMILES FC(C1=C(N2CCC(C3=CC=CC=C3)CC2)C=CN4C1=NN=C4CC5CC5)(F)F
Molecular Weight (g/mol) 400.44
Formula Weight 400.44
Percent Purity 99.24%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation