Learn More
JNJ-42041935, MedChemExpress
Product Code. 16485378
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16485378
Brand: Medchem Express HY12832
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress JNJ-42041935 is a potent, competitive and selective inhibitor of prolyl hydroxylase PHD; inhibits PHD1, PHD2, and PHD3 with pKi values of 7.91±0.04, 7.29 ±0.05, and 7.65±0.09, respectively.
JNJ-42041935 is a potent, competitive and selective inhibitor of prolyl hydroxylase PHD
Specifications
Chemical Name or Material JNJ-42041935
CAS 1193383-09-3
Color White
Purity Grade Notes Research
Molecular Formula C12H6ClF3N4O3
Quantity 10 mg
Solubility Information DMSO : ≥ 36 mg/mL (103.85 mM)
SMILES OC(C(C=N1)=CN1C2=NC3=CC(Cl)=C(OC(F)(F)F)C=C3N2)=O
Molecular Weight (g/mol) 346.65
Formula Weight 346.65
Percent Purity 98.49%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.