missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JMV 2959 hydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
Description
JMV 2959 hydrochloride is a growth hormone secretagogue receptor type 1a (GHS-R1a) antagonist with an IC50 of 32±3 nM in LLC-PK1 cells.
Specifications
Specifications
| Chemical Name or Material | JMV 2959 hydrochloride |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C30H33ClN6O2 |
| Quantity | 25 mg |
| Solubility Information | DMSO : 250 mg/mL (458.65 mM; Need ultrasonic) ∣H2O : 25 mg/mL (45.86 mM; Need ultrasonic) |
| SMILES | O=C(N[C@@H](C1=NN=C(CCC2=CC=CC=C2)N1CC3=CC=C(OC)C=C3)CC4=CNC5=C4C=CC=C5)CN.[H]Cl |
| Molecular Weight (g/mol) | 545.08 |
| Formula Weight | 545.08 |
| Percent Purity | 97.33% |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction