missing translation for 'onlineSavingsMsg'
Learn More

JMV 2959 hydrochloride, MedChemExpress

Product Code. 16470439
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16470439

Brand: Medchem Express HYU00433A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress JMV 2959 hydrochloride is a growth hormone secretagogue receptor type 1a (GHS-R1a) antagonist with an IC50 of 32±3 nM in LLC-PK1 cells.

JMV 2959 hydrochloride is a growth hormone secretagogue receptor type 1a (GHS-R1a) antagonist with an IC50 of 32±3 nM in LLC-PK1 cells.

Specifications

Chemical Name or Material JMV 2959 hydrochloride
Color Off-White
Purity Grade Notes Research
Molecular Formula C30H33ClN6O2
Quantity 10 mg
Solubility Information DMSO : 250 mg/mL (458.65 mM; Need ultrasonic) ∣H2O : 25 mg/mL (45.86 mM; Need ultrasonic)
SMILES O=C(N[C@@H](C1=NN=C(CCC2=CC=CC=C2)N1CC3=CC=C(OC)C=C3)CC4=CNC5=C4C=CC=C5)CN.[H]Cl
Molecular Weight (g/mol) 545.08
Formula Weight 545.08
Percent Purity 97.33%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.