Learn More
Invitrogen™ JMJD1B Recombinant Rabbit Monoclonal Antibody (PSH0-11)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA545972
Description
Predicted band size: 191 kDa Positive Control: Hela cell lysate, NIH/3T3 cell lysate, Hela. Subcellular Location: Nucleus. Sequence Similarities: 92.96% Mouse; 90.14% Rat. Tissue Specificity: Ubiquitous. Highly expressed in placenta, skeletal muscle, kidney, heart and liver.
Histone demethylase that specifically demethylates 'Lys-9' of histone H3, thereby playing a central role in histone code. Demethylation of Lys residue generates formaldehyde and succinate. May have tumor suppressor activity.
Specifications
| JMJD1B | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide | |
| Q6ZPY7, Q7LBC6 | |
| KDM3B | |
| Recombinant protein within human KDM3B aa 251-500. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| ChIP Assay, Western Blot, Immunocytochemistry | |
| PSH0-11 | |
| Unconjugated | |
| KDM3B | |
| 5830462I21Rik; 5qNCA; C5orf7; JHDM2B; JmjC domain-containing histone demethylation protein 2B; JMJD1B; jumonji domain containing 1B; jumonji domain-containing protein 1B; KDM3B; KDM3B lysine (K)-specific demethylase 3B; KIAA1082; lysine (K)-specific demethylase 3B; lysine demethylase 3B; lysine-specific demethylase 3B; mKIAA1082; NET22; Nuclear protein 5qNCA | |
| Rabbit | |
| Protein A | |
| RUO | |
| 277250, 51780 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.