Learn More
JH-II-127, TRC
Specifications
Specifications
| Chemical Name or Material | JH-II-127 |
| CAS | 1700693-08-8 |
| Molecular Formula | C19H21ClN6O3 |
| Quantity | 5 mg |
| Synonym | [4-[[5-Chloro-4-(methylamino)-7H-pyrrolo[2,3-d]pyrimidin-2-yl]amino]-3-methoxyphenyl]-4-morpholinyl-methanone |
| InChI Formula | InChI=1S/C19H21ClN6O3/c1-21-16-15-12(20)10-22-17(15)25-19(24-16)23-13-4-3-11(9-14(13)28-2)18(27)26-5-7-29-8-6-26/h3-4,9-10H,5-8H2,1-2H3,(H3,21,22,23,24,25) |
| SMILES | O=C(C1=CC(OC)=C(C=C1)NC2=NC(NC)=C3C(Cl)=CNC3=N2)N4CCOCC4 |
| IUPAC Name | [4-[[5-chloro-4-(methylamino)-7H-pyrrolo[2,3-d]pyrimidin-2-yl]amino]-3-methoxyphenyl]-morpholin-4-ylmethanone |
| Molecular Weight (g/mol) | 416.87 |
| Formula Weight | 416.1364 |