missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Japanese Encephalitis Virus Core Protein C Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5117304

Product Code. 17239803

  • 506.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Centrifuge briefly prior to opening.

Japanese encephalitis (JE) is an infection of the brain caused by the Japanese encephalitis virus (JEV). While most infections result in little or no symptoms, occasional inflammation of the brain occurs. In these cases, symptoms may include headache, vomiting, fever, confusion and seizures. Nuclear localization of Japanese encephalitis virus core protein enhances viral replication.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Japanese Encephalitis Virus Core Protein C
Polyclonal
Unconjugated
Japanese Encephalitis
Full length Japanese encephalitis virus Core protein C Recombinant protein (Japanese Encephalitis Virus strain Jaoars982).
100 μL
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.02 mg/mL
PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Rabbit
Antigen affinity chromatography
RUO
Virus
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.