missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Jak3 Mouse anti-Human, Alexa Fluor 647, Clone: 452524, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems IC46991R-100UG
This item is not returnable.
View return policy
Description
Jak3 Monoclonal specifically detects Jak3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| Jak3 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| EC 2.7.10, JAK3_HUMAN, JAK-3EC 2.7.10.2, JAKL, JAKLtyrosine-protein kinase JAK3, Janus kinase 3Leukocyte janus kinase, LJAK, L-JAKJanus kinase 3 (a protein tyrosine kinase, leukocyte) | |
| E. coli-derived recombinant human Jak3 Gly46-Thr209 Accession # P52333 | |
| RUO | |
| 3718 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 452524 | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P52333 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Jak3 in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only (RUO)