missing translation for 'onlineSavingsMsg'
Learn More

IWP-O1, MedChemExpress

Product Code. p-200096787
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16425927 5 mg 5mg
16424476 100 mg 100mg
16412666 25 mg 25mg
16409746 50 mg 50mg
16403198 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16425927 Supplier Medchem Express Supplier No. HY100853

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress IWP-O1 is a highly potent Porcupine (Porcn) inhibitor, with an EC50 of 80 pM in L-Wnt-STF cells. IWP-O1 prevents the secretion of Wnt proteins. IWP-O1 suppresses the phosphorylation of Dvl2/3 and LRP6 in HeLa cells.

IWP-O1 is a highly potent Porcupine (Porcn) inhibitor, with an EC50 of 80 pM in L-Wnt-STF cells. IWP-O1 prevents the secretion of Wnt proteins. IWP-O1 suppresses the phosphorylation of Dvl2/3 and LRP6 in HeLa cells.

Specifications

Chemical Name or Material IWP-O1
CAS 2074607-48-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C26H20N6O
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (289.03 mM; Need ultrasonic)
SMILES O=C(CN1N=NC(C2=CC=NC=C2)=C1C3=CC=CC=C3)NC4=NC=C(C5=CC=CC=C5)C=C4
Molecular Weight (g/mol) 432.48
Formula Weight 432.48
Percent Purity 99.61%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.