Learn More
IWP-O1, MedChemExpress
Product Code. 16425927
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16425927
Brand: Medchem Express HY100853
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress IWP-O1 is a highly potent Porcupine (Porcn) inhibitor, with an EC50 of 80 pM in L-Wnt-STF cells. IWP-O1 prevents the secretion of Wnt proteins. IWP-O1 suppresses the phosphorylation of Dvl2/3 and LRP6 in HeLa cells.
IWP-O1 is a highly potent Porcupine (Porcn) inhibitor, with an EC50 of 80 pM in L-Wnt-STF cells. IWP-O1 prevents the secretion of Wnt proteins. IWP-O1 suppresses the phosphorylation of Dvl2/3 and LRP6 in HeLa cells.
Specifications
Chemical Name or Material IWP-O1
CAS 2074607-48-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C26H20N6O
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (289.03 mM; Need ultrasonic)
SMILES O=C(CN1N=NC(C2=CC=NC=C2)=C1C3=CC=CC=C3)NC4=NC=C(C5=CC=CC=C5)C=C4
Molecular Weight (g/mol) 432.48
Formula Weight 432.48
Percent Purity 99.61%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease