missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITX3, MedChemExpress
Description
ITX3 is a specific and nontoxic inhibitor of the TrioN (N-terminal GEF domain of the multidomain Trio protein) with IC50 of 76 uM
Specifications
Specifications
| Chemical Name or Material | ITX3 |
| CAS | 347323-96-0 |
| Color | Light Green |
| Purity Grade Notes | Research |
| Molecular Formula | C22H17N3OS |
| Quantity | 50 mg |
| Solubility Information | DMSO : 2 mg/mL (5.38 mM; ultrasonic and warming and heat to 80°C) |
| SMILES | O=C(/C1=C\C2=C(C)N(C(C)=C2)C3=CC=CC=C3)N4C(S1)=NC5=CC=CC=C45 |
| Molecular Weight (g/mol) | 371.45 |
| Formula Weight | 371.45 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?