missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ITPKB Polyclonal antibody specifically detects ITPKB in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
Specifications
| Antigen | ITPKB |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 525 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 2.7.1.127, inositol 14,5-trisphosphate 3-kinase BIP3K, inositol-trisphosphate 3-kinase B, InsP 3-kinase B, IP3 3-kinase B, IP3-3KB, IP3K B, IP3KB, IP3K-B, PIG37, proliferation-inducing protein 37 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human ITPKB (NP_002212.3).,, Sequence:, MAVYCYALNSLVIMNSANEMKSGGGPGPSGSETPPPPRRAVLSPGSVFSPGRGASFLFPPAESLSPEEPRSPGGWRSGRRRLNSSSGSGSGSSGSSVSSPSWAGRLRGDRQQVVAAGTLSPPGPEEAKRKLRILQRELQNVQVNQKVGMFEAHIQAQSSAIQAPRSPRLGRARSPSPCPFRSSSQPPGRVLVQGARSEERRTKSWGEQCPETSGTDSGRKGGPSLCSSQVKKGMPPLPGRAAPTGSEAQGPSAFVRMEKG |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?