missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ITGA11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103443
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ITGA11.
This gene encodes an alpha integrin. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This protein contains an I domain, is expressed in muscle tissue, dimerizes with beta 1 integrin in vitro, and appears to bind collagen in this form. Therefore, the protein may be involved in attaching muscle tissue to the extracellular matrix. Alternative transcriptional splice variants have been found for this gene, but their biological validity is not determined.
Specifications
| ITGA11 | |
| Polyclonal | |
| Unconjugated | |
| Itga11 | |
| 4732459H24Rik; HsT18964; integrin a11; integrin alpha 11; integrin alpha-11; integrin subunit alpha 11; integrin, alpha 11; Itga11; MSTP018 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 22801, 319480 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P61622, Q9UKX5 | |
| Itga11 | |
| A synthesized peptide derived from human ITGA11(Accession Q9UKX5), corresponding to amino acid residues R1054-L1074. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction