Learn More
Isosteviol, MedChemExpress
Product Code. 16499148
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499148 10 mg 10mg
16481558 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16499148 Supplier Medchem Express Supplier No. HYN0872
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Isosteviol ((-)-Isosteviol) is a derivative of Stevioside through acid catalyzed hydrolysis of Stevioside. Isosteviol inhibits DNA polymerase and DNA topoisomerase and has antibacterial, anticancer and anti-tuberculosis effects.
Isosteviol ((-)-Isosteviol) is a derivative of Stevioside through acid catalyzed hydrolysis of Stevioside. Isosteviol inhibits DNA polymerase and DNA topoisomerase and has antibacterial, anticancer and anti-tuberculosis effects.
Specifications
Chemical Name or Material Isosteviol
Physical Form Solid
CAS 27975-19-5
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C20H30O3
For Use With (Application) COVID-19-immunoregulation
Synonym (-)-Isosteviol iso-Steviol
Solubility Information DMSO : ≥ 100 mg/mL (314.02 mM)
SMILES C[C@]([C@]1([H])CC2)(CCC[C@@]1(C)C(O)=O)[C@@]3([H])[C@@]2(CC4=O)C[C@]4(C)CC3
Molecular Weight (g/mol) 318.45
Formula Weight 318.45
Percent Purity 98.0%
Grade Research