missing translation for 'onlineSavingsMsg'
Learn More

Isoquinoline-3-carbonitrile, 99%, Thermo Scientific™

Product Code. 11378726
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378726 250 mg 250mg
11368726 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11378726 Supplier Thermo Scientific Alfa Aesar Supplier No. L19426.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Isoquinoline-3-carbonitrile is employed as analytical standard to study the thermal degradation of 2-chlorobenzylidenemalononitrile (CS riot control agent) at elevated temperatures.

Solubility
Insoluble in water.

Notes
Keep the container tightly sealed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26947-41-1
Molecular Formula C10H6N2
Molecular Weight (g/mol) 154.172
MDL Number MFCD00075136
InChI Key GXXBVBVNGMCFIU-UHFFFAOYSA-N
Synonym 3-isoquinolinecarbonitrile, 3-cyanoisoquinoline, 3-cyano-isoquinoline, pubchem16887, acmc-1cit2, 2,4,6-cycloheptartriene-1-carbonitrile
PubChem CID 5076808
IUPAC Name isoquinoline-3-carbonitrile
SMILES C1=CC=C2C=NC(=CC2=C1)C#N

Specifications

Melting Point 125°C to 128°C
Quantity 250 mg
Beilstein 114887
Solubility Information Insoluble in water.
Formula Weight 154.17
Percent Purity 99%
Chemical Name or Material Isoquinoline-3-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.