Learn More
Isopropyl bromoacetate, TRC
Specifications
Specifications
| Chemical Name or Material | Isopropyl bromoacetate |
| CAS | 29921-57-1 |
| Molecular Formula | C5H9BrO2 |
| Quantity | 100 mg |
| Synonym | Propan-2-yl-2-bromoacetate, Acetic acid, 2-bromo-1-methylethyl ester |
| InChI Formula | InChI=1S/C5H9BrO2/c1-4(2)8-5(7)3-6/h4H,3H2,1-2H3 |
| SMILES | CC(C)OC(=O)CBr |
| IUPAC Name | propan-2-yl 2-bromoacetate |
| Molecular Weight (g/mol) | 181.03 |
| Formula Weight | 179.9786 |