Learn More
Isophthaldehyde, TRC
Specifications
Specifications
| Chemical Name or Material | Isophthaldehyde |
| CAS | 626-19-7 |
| Molecular Formula | C8H6O2 |
| Quantity | 250 g |
| Synonym | 1,3-Benzenedicarboxaldehyde, 1,3-Benzenedialdehyde, 1,3-Diformylbenzene, 3-Formylbenzaldehyde, Benzene-1,3-dicarbaldehyde, Isophthaldialdehyde, NSC 5092, m-Benzenedialdehyde, m-Benzenedicarbaldehyde, m-Benzenedicarboxaldehyde, m-Diformylbenzene, m-Formylbenzaldehyde, m-Phthalaldehyde |
| InChI Formula | InChI=1S/C8H6O2/c9-5-7-2-1-3-8(4-7)6-10/h1-6H |
| SMILES | O=Cc1cccc(C=O)c1 |
| IUPAC Name | benzene-1,3-dicarbaldehyde |
| Molecular Weight (g/mol) | 134.13 |
| Formula Weight | 134.0368 |