Learn More
Isonitrosoacetophenone, 97%, Thermo Scientific™
Product Code. 11371507
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11371507 10 g 10g
11381507 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11371507 Supplier Thermo Scientific Alfa Aesar Supplier No. L02098.09
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 532-54-7
Molecular Formula C8H7NO2
Molecular Weight (g/mol) 149.149
MDL Number MFCD00002122
InChI Key MLNKXLRYCLKJSS-RMKNXTFCSA-N
Synonym isonitrosoacetophenone, 2-isonitrosoacetophenone, benzoylformaldoxime, inaf, phenylglyoxal aldoxime, phenylglyoxal 2-oxime, 2-hydroxyiminoacetophenone, phenylglyoxal monoxime, acetophenone, 2-hydroxyimino, phenylglyoxaldoxime
PubChem CID 9566037
IUPAC Name (2E)-2-hydroxyimino-1-phenylethanone
SMILES C1=CC=C(C=C1)C(=O)C=NO
Specifications
Melting Point 124°C to 128°C
Quantity 10 g
Beilstein 2041691
Merck Index 14,5191
Formula Weight 149.15
Percent Purity 97%
Chemical Name or Material Isonitrosoacetophenone