Learn More
Isoconazole, TRC
Specifications
Specifications
| Chemical Name or Material | Isoconazole |
| CAS | 27523-40-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C18 H14 Cl4 N2 O |
| Quantity | 10 mg |
| Synonym | Imidazole, 1-[2,4-dichloro-β-[(2,6-dichlorobenzyl)oxy]phenethyl]- (8CI), 1-[2-(2,4-Dichlorophenyl)-2-[(2,6-dichlorophenyl)methoxy]ethyl]-1H-imidazole, (±)-Isconazole, Isoconazole |
| InChI Formula | InChI=1S/C18H14Cl4N2O/c19-12-4-5-13(17(22)8-12)18(9-24-7-6-23-11-24)25-10-14-15(20)2-1-3-16(14)21/h1-8,11,18H,9-10H2 |
| SMILES | Clc1ccc(C(Cn2ccnc2)OCc3c(Cl)cccc3Cl)c(Cl)c1 |
| IUPAC Name | 1-[2-(2,4-dichlorophenyl)-2-[(2,6-dichlorophenyl)methoxy]ethyl]imidazole |
| Molecular Weight (g/mol) | 416.13 |
| Show More |