Learn More
Isoconazole nitrate, MedChemExpress
Product Code. 16474597
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Product Code. 16474597
Brand: Medchem Express HYB1444
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Isoconazole nitrate is a broad-spectrum antimicrobial agent with a highly effective antimycotic and gram-positive antibacterial activity, exhibiting a rapid rate of absorption and low systemic exposure potential.
Isoconazole nitrate is a broad-spectrum antimicrobial agent with a highly effective antimycotic and gram-positive antibacterial activity, exhibiting a rapid rate of absorption and low systemic exposure potential.
Specifications
Chemical Name or Material Isoconazole nitrate
Physical Form Solid
CAS 24168-96-5
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C18H15Cl4N3O4
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 100 mg/mL (208.71 mM; Need ultrasonic)
SMILES ClC1=C(COC(C2=CC=C(Cl)C=C2Cl)CN3C=CN=C3)C(Cl)=CC=C1.O[N+]([O-])=O
Molecular Weight (g/mol) 479.14
Formula Weight 479.14
Percent Purity 98.0%
Grade Research