missing translation for 'onlineSavingsMsg'
Learn More

Isoacteoside, MedChemExpress

Product Code. p-200099390
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16488956

Brand: Medchem Express HYN0022

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Isoacteoside is a natural compound which exhibit significant inhibition of advanced glycation end product formation with IC50 values of 4.6-25.7 μM, compared with those of aminoguanidine (IC50=1,056 μM) and quercetin (IC50=28.4 μM) as positive controls.

Isoacteoside is a natural compound which exhibit significant inhibition of advanced glycation end product formation with IC50 values of 4.6-25.7 μM, compared with those of aminoguanidine (IC50=1,056 μM) and quercetin (IC50=28.4 μM) as positive controls.

Specifications

Chemical Name or Material Isoacteoside
CAS 61303-13-7
Color White
Purity Grade Notes Research
Molecular Formula C29H36O15
Quantity 100 mg
Synonym Isoverbascoside
Solubility Information DMSO : 250 mg/mL (400.26 mM; Need ultrasonic)
SMILES OC1=CC(/C=C/C(OC[C@@H]2[C@@H](O)[C@H](O[C@@H]3O[C@@H](C)[C@H](O)[C@@H](O)[C@H]3O)[C@@H](O)[C@H](OCCC4=CC=C(O)C(O)=C4)O2)=O)=CC=C1O
Molecular Weight (g/mol) 624.59
Formula Weight 624.59
Percent Purity 99.13%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.