missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IRX2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16623515
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16623515 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16623515 Supplier Invitrogen™ Supplier No. PA599262

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total IRX2.

IRX2 is a member of the Iroquois homeobox gene family. Members of this family appear to play multiple roles during pattern formation of vertebrate embryos.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRX2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IRX2
Gene Accession No. P81066, Q9BZI1
Gene Alias homeodomain protein IRXA2; iroquois homeobox 2; iroquois homeobox protein 2; Iroquois related homeobox 2 (Drosophila); iroquois-class homeobox protein Irx6; Iroquois-class homeodomain protein IRX-2; Irx2; Irx6; IRXA2
Gene Symbols IRX2
Host Species Rabbit
Immunogen A synthesized peptide derived from human IRX2(Accession Q9BZI1), corresponding to amino acid residues D237-P287.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 153572, 16372, 306657
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.