missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRSp53 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11087-2-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene has been identified as a brain-specific angiogenesis inhibitor (BAI1)-binding protein. This adaptor protein links membrane bound G-proteins to cytoplasmic effector proteins. This protein functions as an insulin receptor tyrosine kinase substrate and suggests a role for insulin in the central nervous system. It also associates with a downstream effector of Rho small G proteins, which is associated with the formation of stress fibers and cytokinesis. This protein is involved in lamellipodia and filopodia formation in motile cells and may affect neuronal growth-cone guidance. This protein has also been identified as interacting with the dentatorubral-pallidoluysian atrophy gene, which is associated with an autosomal domit neurodegenerative disease. Alternative splicing results in multiple transcript variants encoding distinct isoforms.Specifications
| IRSp53 | |
| Polyclonal | |
| Unconjugated | |
| BAIAP2 | |
| BAI associated protein 2, BAI1 associated protein 2, BAIAP2, BAP2, Fas ligand associated factor 3, FLAF3, Insulin receptor substrate p53, IRS 58, IRSp53, IRSp53/58, Protein BAP2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10458, 108100, 117542 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6GMN2, Q8BKX1, Q9UQB8 | |
| BAIAP2 | |
| IRSp53 Fusion Protein Ag1552 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction