Learn More
Iron(II) Fumarate, TRC
Product Code. 30134704
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134704 25 g 25g
30135577 5 g 5g
30010893 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134704 Supplier Toronto Research Chemicals Supplier No. TRCI77530025G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 141-01-5
Molecular Formula C4 H2 O4 . Fe
Molecular Weight (g/mol) 169.9
Synonym Ferrous fumarate, Iron(II) (E)-butenedioate
IUPAC Name (E)-but-2-enedioate;iron(2+)
SMILES [Fe+2].[O-]C(=O)\C=C\C(=O)[O-]
Specifications
Quantity 25 g
InChI Formula InChI=1S/C4H4O4.Fe/c5-3(6)1-2-4(7)8;/h1-2H,(H,5,6)(H,7,8);/q;+2/p-2/b2-1+;
Formula Weight 169.9303
Chemical Name or Material Iron(II) Fumarate