missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF3 Antibody (3F10), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04308H
This item is not returnable.
View return policy
Description
IRF3 Monoclonal antibody specifically detects IRF3 in Human samples. It is validated for Western Blot, ELISA, KnockDown
Specifications
| IRF3 | |
| Monoclonal | |
| HRP | |
| interferon regulatory factor 3, IRF-3 | |
| Recombinant human IRF3 (108-166aa) purified from E. coli | |
| 0.1 mL | |
| Immunology, Innate Immunity, Transcription Factors and Regulators | |
| 3661 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, KnockDown | |
| 3F10 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction