missing translation for 'onlineSavingsMsg'
Learn More

IRAK-1-4 Inhibitor I, MedChemExpress

Product Code. 16438347
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16438347

Brand: Medchem Express HY13329

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress IRAK-1-4 Inhibitor I is an inhibitor of interleukin-1 receptor-associated kinase 1/4 (IRAK 1/4) with IC50s of 0.2 μM and 0.3 μM, respectively.

IRAK-1-4 Inhibitor I is an inhibitor of interleukin-1 receptor-associated kinase 1/4 (IRAK 1/4) with IC50s of 0.2 μM and 0.3 μM, respectively.

Specifications

Chemical Name or Material IRAK-1-4 Inhibitor I
CAS 509093-47-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H21N5O4
Quantity 5 mg
Synonym IRAK-1/4 Inhibitor I
Solubility Information DMF : 25 mg/mL (63.23 mM; ultrasonic and warming and heat to 60°C) ∣DMSO : 5 mg/mL (12.65 mM; ultrasonic and adjust pH to 3 with HCl) ∣Ethanol : < 1 mg/mL (insoluble) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=CC=CC([N+]([O-])=O)=C1)NC2=NC3=CC=CC=C3N2CCN4CCOCC4
Molecular Weight (g/mol) 395.41
Formula Weight 395.41
Percent Purity 99.2%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.