missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IQCB1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15573881
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15573881 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15573881 Supplier Invitrogen™ Supplier No. PA536484

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 69 kDa. Purity is >95% by SDS-PAGE.

This gene was first characterized as part of a cluster of genes located within the human major histocompatibility complex class III region. This gene encodes a nuclear protein that is cleaved by caspase 3 and is implicated in the control of apoptosis. In addition, the protein forms a complex with E1A binding protein p300 and is required for the acetylation of p53 in response to DNA damage. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IQCB1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Iqcb1
Gene Accession No. Q15051, Q8BP00
Gene Alias 6820449I09Rik; AV128382; IQ calmodulin-binding motif containing 1; IQ calmodulin-binding motif-containing protein 1; IQ motif containing B1; IQCB1; KIAA0036; nephrocystin 5; Nephrocystin-5; NPHP5; OK/SW-cl.85; p53 and DNA damage-regulated IQ motif protein; PIQ; SLSN5
Gene Symbols Iqcb1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 432-486 of human Nephrocystin-5.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303915, 320299, 9657
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.