missing translation for 'onlineSavingsMsg'
Learn More

IQ-1S free acid, MedChemExpress

Product Code. p-200097226
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16484266

Brand: Medchem Express HY100233

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress IQ-1S free acid is a prospective inhibitor of NF-κB/activating protein 1 (AP-1) activity with an IC50 of 2.3±0.41 μM. IQ-1S free acid has binding affinity (Kd values) in the nanomolar range for all three JNKs with Kds of 100 nM, 240 nM, and 360 nM for JNK3, JNK1, and JNK2, respectively.

IQ-1S free acid is a prospective inhibitor of NF-κB/activating protein 1 (AP-1) activity with an IC50 of 2.3±0.41 μM. IQ-1S free acid has binding affinity (Kd values) in the nanomolar range for all three JNKs with Kds of 100 nM, 240 nM, and 360 nM for JNK3, JNK1, and JNK2, respectively.

Specifications

Chemical Name or Material IQ-1S free acid
Physical Form Solid
CAS 23146-22-7
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C15H9N3O
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 32 mg/mL (129.42 mM)
SMILES O/N=C1C2=C(C3=NC4=CC=CC=C4N=C3\1)C=CC=C2
Molecular Weight (g/mol) 247.25
Formula Weight 247.25
Percent Purity 99.35%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.