Learn More
IQ-1S free acid, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Description
IQ-1S free acid is a prospective inhibitor of NF-κB/activating protein 1 (AP-1) activity with an IC50 of 2.3±0.41 μM. IQ-1S free acid has binding affinity (Kd values) in the nanomolar range for all three JNKs with Kds of 100 nM, 240 nM, and 360 nM for JNK3, JNK1, and JNK2, respectively.
Specifications
Specifications
| Chemical Name or Material | IQ-1S free acid |
| Physical Form | Solid |
| CAS | 23146-22-7 |
| Color | White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C15H9N3O |
| For Use With (Application) | Cancer-Kinase/protease |
| Solubility Information | DMSO : ≥ 32 mg/mL (129.42 mM) |
| SMILES | O/N=C1C2=C(C3=NC4=CC=CC=C4N=C3\1)C=CC=C2 |
| Show More |