missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IPO5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13434617
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13434617 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13434617 Supplier Invitrogen™ Supplier No. PA530076

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: A549, NT2D1, HeLa, Jurkat, Raji, mouse brain. Predicted reactivity: Mouse (98%), Xenopus laevis (81%), Pig (97%), Chimpanzee (100%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Nucleocytoplasmic transport, a signal- and energy-dependent process, takes place through nuclear pore complexes embedded in the nuclear envelope. The import of proteins containing a nuclear localization signal (NLS) requires the NLS import receptor, a heterodimer of importin alpha and beta subunits also known as karyopherins. Importin alpha binds the NLS-containing cargo in the cytoplasm and importin beta docks the complex at the cytoplasmic side of the nuclear pore complex. In the presence of nucleoside triphosphates and the small GTP binding protein Ran, the complex moves into the nuclear pore complex and the importin subunits dissociate. Importin alpha enters the nucleoplasm with its passenger protein and importin beta remains at the pore. Interactions between importin beta and the FG repeats of nucleoporins are essential in translocation through the pore complex. The protein encoded by this gene is a member of the importin beta family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IPO5
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.78 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene IPO5
Gene Accession No. O00410, Q8BKC5
Gene Alias 1110011C18Rik; 5730478E03Rik; AA409333; C76941; IMB3; imp5; importin 5; importin beta-3 subunit; importin subunit beta-3; Importin-5; IPO 5; IPO5; karyopherin (importin) beta 3; Karyopherin beta 3; karyopherin beta-3; Kpnb3; MGC2068; Pse1; RAN binding protein 5; Ran_GTP binding protein 5; ran-binding protein 5; RanBP 5; Ranbp5
Gene Symbols IPO5
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human IPO5. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3843, 70572
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.