missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ipatasertib, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
1 g
Unit Size:
100mg
10mg
1g
200mg
500mg
50mg
5mg
Description
Ipatasertib (GDC-0068) is a highly selective and ATP-competitive pan-Akt inhibitor with IC50s of 5, 18 and 8 nM for Akt1, Akt2 and Akt3, respectively.
Specifications
Specifications
| Chemical Name or Material | Ipatasertib |
| Physical Form | Powder |
| CAS | 1001264-89-6 |
| Color | Off-White |
| Quantity | 200 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C24H32ClN5O2 |
| For Use With (Application) | Cancer-Kinase/protease |
| Synonym | GDC-0068 RG7440 |
| Solubility Information | DMSO : 220 mg/mL (480.35 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction